missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MID1IP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519370
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
Specifications
| MID1IP1 | |
| Polyclonal | |
| Unconjugated | |
| MID1IP1 | |
| 3110038L01Rik; FLJ10386; G12-like; gastrulation specific G12 homolog; gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1; MID1 interacting protein 1 (gastrulation specific G12 homolog); Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)); MID1 interacting protein 1 (gastrulation specific G12-like); mid1-interacting G12-like protein; Mid1-interacting protein 1; MID1IP1; mid1ip1 {ECO:0000250; Mig12; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; slap; spot 14-like androgen-inducible protein; spot 14-R; spot 14-related protein; STRAIT11499; THRSPL; UniProtKB:Q9CQ20} | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 58526 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9NPA3 | |
| MID1IP1 | |
| Synthetic peptide sequence (HTVLSKLTRK) corresponding to the internal amino acids of MID1IP1 (aa 155-165). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction