missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MID1IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568224
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.
Specifications
| MID1IP1 | |
| Polyclonal | |
| Unconjugated | |
| MID1IP1 | |
| 3110038L01Rik; FLJ10386; G12-like; gastrulation specific G12 homolog; gastrulation-specific G12-like protein; MID1 interacting G12-like protein; MID1 interacting protein 1; MID1 interacting protein 1 (gastrulation specific G12 homolog); Mid1 interacting protein 1 (gastrulation specific G12-like (zebrafish)); MID1 interacting protein 1 (gastrulation specific G12-like); mid1-interacting G12-like protein; Mid1-interacting protein 1; MID1IP1; mid1ip1 {ECO:0000250; Mig12; Protein STRAIT11499; Protein STRAIT11499 homolog; S14R; slap; spot 14-like androgen-inducible protein; spot 14-R; spot 14-related protein; STRAIT11499; THRSPL; UniProtKB:Q9CQ20} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 404280, 58526, 68041 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6P7D5, Q9CQ20, Q9NPA3 | |
| MID1IP1 | |
| Synthesized peptide derived from human MID1IP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction