missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID1IP1, Mouse anti-Human, Clone: 8G8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00058526-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MMQICDTYNQKHSLFNAMNRFIGAVNNMDQTVMVPSLLRDVPLADPGLDNDVGVEVGGSGGCLEERTPPVPDSGSANGSFFAPSRDMYSHYVLLKSIRNDIEWGVLHQPPPPAGSEEGSAWKSKDILVDLGHLEGADAGEEDLEQQFHYHLRGLHTVLSKLTRKANILTNRYKQEIGFGNWGHSpecifications
| MID1IP1 | |
| Monoclonal | |
| Unconjugated | |
| MID1 interacting protein 1 (gastrulation specific G12 homolog (zebrafish)) | |
| FLJ10386/G12-like/MIG12/STRAIT11499/THRSPL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 8G8 | |
| In 1x PBS, pH 7.4 | |
| NM_021242.3 | |
| MID1IP1 | |
| MID1IP1 (NP_067065.1, 1 a.a. ∼ 183 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 58526 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction