Learn More
MID1IP1 Antibody (079), mFluor Violet 610 SE, Novus Biologicals™
Product Code. 18401889 Shop All Bio Techne Products
Product Code. 18401889 Supplier Novus Biologicals Supplier No. NBP290265MFV610
Rabbit Monoclonal Antibody
MID1IP1 Monoclonal antibody specifically detects MID1IP1 in Human samples. It is validated for ELISA