missing translation for 'onlineSavingsMsg'
Learn More

MID1IP1 Antibody (005), DyLight 488, Novus Biologicals™

Product Code. 18389850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389850 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389850 Supplier Novus Biologicals Supplier No. NBP290264G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MID1IP1 Monoclonal specifically detects MID1IP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MID1IP1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 5
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias FLJ10386, G12-like, Gastrulation-specific G12-like protein, MID1 interacting protein 1 (gastrulation specific G12 homolog (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like (zebrafish)), MID1 interacting protein 1 (gastrulation specific G12-like), Mid1-interacting G12-like protein, mid1-interacting protein 1, MIG12MID1 interacting G12-like protein, Protein STRAIT11499, S14R, Spot 14-R, Spot 14-related protein, STRAIT11499, THRSPL
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human MID1IP1 (Q9NPA3; Met1-His183).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58526
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.