missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£174.00 - £395.00
Specifications
| Antigen | MID1 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18363644
|
Novus Biologicals
NBP3-05061-20UL |
20 μL |
£174.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099343
|
Novus Biologicals
NBP3-05061-100UL |
100 μL |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MID1 Polyclonal specifically detects MID1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MID1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 6.3.2.-, FLJ57031, FLJ58683, FLJ76288, FXYGBBB1, Midin, midline 1 (Opitz/BBB syndrome), RNF59MIDIN, TRIM18mouse, homolog of | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 478-667 of human MID1 (NP_000372.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Zinc Finger | |
| PBS with 50% glycerol, pH7.3. | |
| 4281 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title