missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MID1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536305
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 75 kDa. Purity is >95% by SDS-PAGE.
Midline-1 (Tripartite motif-containing protein 18, Putative transcription factor XPRF, RING finger protein 59) is a 667 amino acid protein encoded by the human gene MID1. Midline-1 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-1 is believed to have E3 ubiquitin ligase activity which targets the catalytic subunit of protein phosphatase 2 for degradation. It is a cytoplasmic protein found as a homodimer or heterodimer with Midline-2. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Defects in MID1 are the cause of Opitz syndrome type I (OS-I). OS-I is an X-linked recessive disorder characterized by hypertelorism, genital-urinary defects such as hypospadias in males and splayed labia in females, lip-palate-laryngotracheal clefts, imperforate anus, developmental delay and congenital heart defects.
Specifications
| MID1 | |
| Polyclonal | |
| Unconjugated | |
| MID1 | |
| 61B3-R; BBBG1; DXHXS1141; E3 ubiquitin-protein ligase Midline-1; Finger on X and Y (in rat only on X); Fxy; GBBB1; Mid1; Midin; midline 1; midline 1 (Opitz/BBB syndrome); midline 1 RING finger protein; Midline1; midline-1; OGS1; Opitz/BBB syndrome; OS; OSX; Putative transcription factor XPRF; RING finger protein 59; RING finger protein Midline-1; RING-type E3 ubiquitin transferase Midline-1; RNF59; TRIM18; tripartite motif protein TRIM18; tripartite motif-containing protein 18; XPRF; zinc finger on X and Y, mouse, homolog of; ZNFXY | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17318, 4281, 54252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15344, O70583, P82458 | |
| MID1 | |
| Synthetic peptide corresponding to amino acids 71-120 of human Midline-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction