missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICS1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519249
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
Specifications
| MICS1 | |
| Polyclonal | |
| Unconjugated | |
| GHITM | |
| 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 27069 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H3K2 | |
| GHITM | |
| Synthetic peptide sequence (PSREYATKTRIGIR) corresponding to the internal amino acids of GHITM (aa 41-55). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction