missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563978
This item is not returnable.
View return policy
Description
Immunogen sequence: TKNQWLLTPS REYATKTRIG IRRGRTGQEL KEAALEPSME KIFKIDQMGR W Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
Specifications
| MICS1 | |
| Polyclonal | |
| Unconjugated | |
| GHITM | |
| 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3K2 | |
| GHITM | |
| Recombinant protein corresponding to Human MICS1. Recombinant protein control fragment (Product #RP-104182). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction