Learn More
Corning™ Microvolume Pipet Tips, Sterile
Provide accurate, reliable performance in the stated volume range
Brand:  Corning™ 4830
Code : 66
Additional Details : Weight : 1.13398kg
 View more versions of this product
Product Code. 10072521
  • £92.59 / Pack of 960
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Microvolume tips provide accurate, reliable performance in the 0.1 μL to 10 μL range for major brand ultra-micropipettors.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
Specifications
0.5 to 10 μL
Natural
Rack
Rack
Polypropylene
Sterile
DNase, RNase, and Pyrogen free
Microvolume, Round
Eppendorf Pipetter, Universal Fit Ultramicro