Learn More
Ohaus™ Microtube Foam Insert
Shop All Ohaus Products
Click to view available options
Holds:
38 Mircotubes
Unit Size:
Each
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
| Holds | 38 Mircotubes |
| Product Type | Foam Insert |
| Material | Foam |
| Description | Foam Insert (38) 1.5/2.0 mL Microtubes |
| No. of Positions | 38 |