Learn More
Thermo Scientific™ MicroTest™ M4RT 3mL w/o Beads Tube, Red
Description
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.
MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
- Stringent quality control, including cellular toxicity on each lot number
- Patented formulations with stabilizing proteins, buffers, and specific antibiotics
Compatibility and viability
- Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
- Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
Convenient, accommodating format
- 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
- Individual transport media tubes or complete kits offer flexibility and versatility
Specifications
Specifications
| Closure Color | Red |
| Compatibility | Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology |
| Description | M4RT 3 mL Tube without Beads, Red |
| For Use With (Application) | Chlamydiae Viruses |
| Packaging | Packaged in partitioned boxes |
| Quantity | 72 Tubes/Pk |
| Storage Requirements | 2°C to 30°C |
| Transport Temperature | 2° to 30°C |
| Volume (Metric) | 3 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.