missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MicroTest™ Kits

Product Code. 18834220
Change view
Click to view available options
Volume (Metric):
3 mL
Closure Color:
Blue
Lilac
Red
Media Type:
M4
M4-RT
Unit Size:
100 sets
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Closure Color Media Type Unit Size
18834220 3 mL Lilac M4-RT 100 sets
13285519 3 mL Blue M4 Pack of 100
13215549 3 mL Red M4-RT Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18834220 Supplier Thermo Scientific™ Supplier No. SK01360
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maintain viability of viruses chlamydiae, mycoplasma and urealasma during transport and long-term storage with kits including media and traditional swabs.

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and urealasma with quality media, proven collection and transport tools, and safe, reliable packaging. Thermo Scientific™ MicroTest™ Kits come in four media formulations and include two plastic shaft traditional-tipped polyester swabs and a 6 x 9 in. zip-seal bag for safe transport.

Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing

  • Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
  • Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Product Type M4 RT Swab Kit
Format Swab
Media Type M4-RT
Swab Configuration Single Applicator
Closure Color Lilac
Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
Description M4RT (3 mL) NPG Swab Kit, No Beads
For Use With (Application) Chlamydiae Viruses
Quantity 100 Kits/Pk.
Shaft Material Polystrene
Storage Requirements 2°C to 30°C
Tip Type Standard NPG
Volume (Metric) 3 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.