Learn More
Thermo Scientific™ MicroTest™ Kits
Maintain viability of viruses chlamydiae, mycoplasma and urealasma during transport and long-term storage with kits including media and traditional swabs.
Brand: Thermo Scientific™ R12550
Additional Details : Weight : 0.01000kg
Description
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and urealasma with quality media, proven collection and transport tools, and safe, reliable packaging. Thermo Scientific™ MicroTest™ Kits come in four media formulations and include two plastic shaft traditional-tipped polyester swabs and a 6 x 9 in. zip-seal bag for safe transport.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and urealasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas. MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.
MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads, an envelope with two plastic shaft, traditional-tipped polyester swabs, and a 6 x 9 in. zip-seal bag.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
- Stringent quality control, including cellular toxicity on each lot number
- Patented formulations with stabilizing proteins, buffers, and specific antibiotics
- Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
- Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
- 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
- Individual transport media tubes or complete kits offer flexibility and versatility
- Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
- Kits include leak-resistant polypropylene bag for safe sample transport
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
M4 Tube Kit | |
Single Applicator | |
Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology | |
Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas | |
Plastic | |
Traditional |
Tube | |
Blue | |
M4 Kit | |
100 Kits/Pk. | |
-25°C or 2°C to 8°C | |
3 mL |