missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MicroTest™ Flocked Swab Kits

Product Code. 13275539
Change view
Click to view available options
Tip Type:
Traditional
Product Type:
Flocked Swab Kit
Includes:
M4RT Kit w/Traditional-Tipped Flocked Swab
Fill Volume:
3 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Type Product Type Includes Fill Volume Unit Size
13275539 Traditional Flocked Swab Kit M4RT Kit w/Traditional-Tipped Flocked Swab 3 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13275539 Supplier Thermo Scientific™ Supplier No. R12565
  • ONLINE EXCLUSIVE PRICE
    £2020.00
    £1692.36 / Pack of 100
    Save £327.64
    16% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maintain organism viability with kits including quality room-temperature transport media, one traditional-tipped nylon flocked swab and/or one micro-tipped nylon flocked swab.

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging with Thermo Scientific™ MicroTest™ Flocked Swab Kits.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

Microtest Flocked Swab Kits come with 1.5 mL or 3 mL of liquid medium in various tube types, with or without glass beads, individually wrapped swabs, in either a peel pouch format or zip-seal bag. Please refer to the specific product listing for more information about the kit contents.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results

  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics

Compatibility and viability

  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing

Convenient, accommodating format

  • 1.5 mL and 3.0 mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility

Safe, sturdy packaging

  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Tekniske data

Product Type Flocked Swab Kit
Format Tube
Media Type M4-RT
Swab Configuration Single Applicator
Closure Color Red
Compatibility Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
Fill Volume 3 mL
For Use With (Application) Chlamydiae Viruses
Includes M4RT Kit w/Traditional-Tipped Flocked Swab
Quantity 100 Kits/Pk.
Shaft Material Plastic
Storage Requirements 2°C to 30°C
Tip Type Traditional
Transport Temperature 2° to 30°C
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.