missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Purification Combo Kit v2.0, with 8-strips kit

Product Code. 18715475 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18715475

Brand: Applied Biosystems™ A35854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MicroSEQ ID Purification Combo Kit v2.0 includes reagents for post-PCR and post-sequencing clean-up in the MicroSEQ ID microbial identification workflow.

The MicroSEQ ID Purification Combo Kit v2.0 includes reagents for post-PCR and post-sequencing clean-up in the MicroSEQ ID microbial identification workflow. This version contains one tube of ExoSAP-IT Express Reagent for post-PCR clean-up and 12 x 8-strips of MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit for post-sequencing clean-up.

ExoSAP-IT Express Reagent removes unincorporated dNTPs and primers from the post-amplification PCR mix in 5 minutes compared to 30 minutes with the original ExoSAP-IT Reagent while producing equivalent results. The MicroSEQ ID Ultra Sequencing Clean-up kit in 96-well plate or 8-strips format provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions. The end products can be used directly for sequencing on any Applied Biosystems genetic analyzer.

Comparability studies demonstrated equivalent results between the original ExoSAP-IT Reagent and the improved ExoSAP-IT Express Reagent coupled with the MicroSEQ ID Ultra Sequencing Clean-Up kit in the MicroSEQ ID workflow (see figure below). Both ExoSAP-IT and ExoSAP-IT Express enzyme-treated PCR samples produced very similar sequencing results and the correct identification result.

ExoSAP-IT Express Reagent can be used downstream of the following PCR amplification kits:

  • MicroSEQ 500 16S rDNA PCR Kit
  • MicroSEQ Full Gene 16S rDNA PCR Kit
  • MicroSEQD2 rDNA Fungal PCR Kit

MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit can be used downstream of the following sequencing kits:

  • MicroSEQ 500 16S rDNA Sequencing Kit
  • MicroSEQ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ D2 rDNA Fungal Sequencing Kit

Specifications

Content And Storage • 1 x ExoSAP-IT Express Reagent (100 reactions). Store at -5°C to -30°C.
• 1 x MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (12 x 8 strips). Store at 2°C to 8°C.
Format Strip
Sample Type Bacteria, DNA, dsDNA, Fungi
For Use With (Application) Sequencing, PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols
No. of Reactions 100 Reactions
Product Line MicroSEQ™
Quantity 1 kit
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Fluorescent Dye Removal, DNA from Enzymatic Reactions (e.g. PCR)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.