missing translation for 'onlineSavingsMsg'
Learn More

MicroSEQ™ E. coli O157:H7 Detection Kit

Product Code. 12143911
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12143911 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12143911 Supplier Thermo Scientific Oxoid Supplier No. 4427409
  • ONLINE EXCLUSIVE PRICE
    £1025.00
    £928.87 / Pack of 1
    Save £96.13
    9% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance AOAC
Content And Storage This kit contains Assay Beads in 12 8-tube strips (store at 2 to 8°C in the dark), 12 MicroAmp™ Optical 8-Cap Strips, and a tube of Pathogen Detection Negative Control (store at 2 to 8°C in the dark).

There are sufficient reagents to perform 96 tests.
Detectable Analytes E. coli
Detection Method Primer-Probe Detection
Format Kit
Sample Type Any swab type, food, water samples
Type qPCR Detection Kit
Hot Start Built-In Hot Start
Passive Reference Dye ROX (Pre-mixed)
PCR Method qPCR
Polymerase AmpliTaq™ Fast DNA Polymerase
Primer Probe Compatibility TaqMan™
Product Line MicroSEQ
Quantity 96 Tests
Species Human
Strain or Genotype E. coli O157:H7
Target Organism Class E. coli
Test Time Fast
Validated Application Real-Time PCR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.