missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Microsep™ Advance Centrifugal Devices with Omega Membrane

Product Code. 17124751
Change view
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
Includes:
100/Pk.
24/Pk.
Color:
Blue
Clear
Gray
Red
Yellow
Unit Size:
Pack of 100
Pack of 24
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Includes Color Unit Size
17124751 30 kDa 24/Pk. Red Pack of 24
17164741 1 kDa 24/Pk. Yellow Pack of 24
17134751 30 kDa 100/Pk. Red Pack of 100
17194741 3 kDa 100/Pk. Gray Pack of 100
17144751 100 kDa 24/Pk. Clear Pack of 24
17154751 100 kDa 100/Pk. Clear Pack of 100
17114751 10 kDa 100/Pk. Blue Pack of 100
17184741 3 kDa 24/Pk. Gray Pack of 24
17174741 1 kDa 100/Pk. Yellow Pack of 100
17104751 10 kDa 24/Pk. Blue Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17124751 Supplier Cytiva Supplier No. MCP030C41
  • ONLINE EXCLUSIVE PRICE
    £230.37
    £191.00 / Pack of 24
    Save £39.37
    17% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microsep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 5 mL of biological sample.

  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description EFA: 3.3 cm2
Material (Membrane) PES
MWCO 30 kDa
Sterility Non-sterile
Includes 24/Pk.
Volume (Metric) 0.5 to 5 mL (Sample)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.