missing translation for 'onlineSavingsMsg'
Learn More

Carl Zeiss™ AxioCams Microscopy Camera

Product Code. 15293896
Change view
Click to view available options
Description:
AxioCam 105 Color
AxioCam ERc 5s
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15293896 AxioCam ERc 5s Each
15295639 AxioCam 105 Color Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15293896 Supplier Carl Zeiss™ Supplier No. 4265409901000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5 megapixel color microscope camera for documentation in routine labs

  • CMOS microscope camera, 5 megapixels (2560 x 1920)
  • Sensor size of 1/2.5 in. (diagonal 7.1 mm)
  • Pixel size 2.2 x 2.2 μm
  • Color camera
  • Includes ZEN Lite starter software for image capture and making measurements

Specifications

Type Microscopy Camera AxioCam
Description AxioCam ERc 5s
Includes AxioCam ERc 5s Rev.2, driver software 64-bit, connection USB 2.0 Y-cable (2 m), sensor: Micron MT9P031
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.