missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Hazard Warning Tape

Product Code. 10633405
Change view
Click to view available options
Display Text:
Biohazard
Danger Broken Glass
Dimensions (L x W):
66 m x 25 mm
66 m x 50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Dimensions (L x W) unitSize
10633405 Biohazard 66 m x 25 mm Each
12318767 Danger Broken Glass 66 m x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10633405 Supplier Azpack™ Supplier No. 10633405

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Repeats warning with hazard symbol for up to 66m. Azpack™ Hazard Warning Tape is self-adhesive and printed black on a yellow background.

  • Self-adhesive tape
  • Wording and hazard symbol repeated along tape
  • Black on yellow background
  • 66m roll
  • Cardboard core diameter: 75mm
  • Width: 25mm

Specifications

Adhesive Type Self-adhesive
Color Black, Yellow
Format Roll
Material Vinyl
Display Text Biohazard
Quantity 1 Ea.
Dimensions (L x W) 66 m x 25 mm
Length (Metric) 66 m
Width (Metric) 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.