missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Phase Ring for AE2000
Increases image contrast. Motic™ Phase Ring for AE2000 eliminates the phase difference between the background and scattered light rays.
Brand: Motic™ 1101000201641
Code : A0
Additional Details : Weight : 0.01000kg
Description
- For use with AE2000 Binocular Inverted Microscopes
- Use Phase Ring Ph0 (4X) for working in phase contrast with the 4X objective
- Use Phase Ring Ph1 (10X, 20X, 40X) for working in phase contrast with the Ph10X, Ph20X, Ph40X objectives
Specifications
Phase Ring | |
Phase-contrast microscopy | |
Ph 4X objective | |
1 Ea. |