Learn More
Fisherbrand™ Microscope objective S-planachromatic spring loaded accessory for FB69075 - FB69088 100x/1.25/160/0.17 oil
Click to view available options
Magnification Power:
100X
Unit Size:
Each
Description
- Semi-plan achromatic microscope objective
- Accessory for microscope models: FB69075 and FB69088
Specifications
Specifications
| Type | Semi-plan Achromatic Objective |
| Description | 100x/1.25/160/0.17 oil |
| For Use With (Equipment) | Microscope models: FB69075 and FB69088 |
| Magnification Power | 100X |