Learn More
Fisherbrand™ Plan Achromatic Microscope Objective
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Description
- Plan achromatic microscope objective
- Accessory for microscope models: FB69075 and FB69088
Specifications
Specifications
| Type | Plan Achromatic Objective |
| Description | 40x/0.65/160/0.17 PH+ spring loaded |
| For Use With (Equipment) | Microscope models: FB69075 and FB69088 |
| Magnification Power | 40X |