Learn More
Fisherbrand™ Plan Achromatic Microscope Objective
Click to view available options
Magnification Power:
20X / 0.40 / 160 / 0.17
Unit Size:
Each
Description
- For use with compound biological microscopes
- Spring loaded
- 20X / 0.40 / 160 / 0.17
Specifications
Specifications
| Description | Spring loaded |
| For Use With (Equipment) | Compound biological microscope |
| Magnification Power | 20X / 0.40 / 160 / 0.17 |