Learn More
Fisherbrand™ Plan Achromatic Microscope Objective
Product Code. 12348568
Click to view available options
Magnification Power:
10X
Unit Size:
Each
Product Code. 12348568
Brand: Fisherbrand™ 2624.0109
Use this objective with compound biological microscopes. Fisherbrand™ Plan Achromatic Microscope Objectives are ergonomically designed for everyday use.
  • Plan achromatic microscope objective
  • Accessory for microscope models: FB69075 and FB69088
Specifications
Type Plan Achromatic Objective
Description 10x/0.25/160/0.17 PH+ spring loaded
For Use With (Equipment) Microscope models: FB69075 and FB69088
Magnification Power 10X