Learn More
Fisherbrand™ Plan Achromatic Microscope Objective
Click to view available options
Magnification Power:
100X
Unit Size:
Each
Description
- Plan achromatic microscope objective
- Accessory for microscope models: FB69075 and FB69088
Specifications
Specifications
| Type | Plan Achromatic Objective |
| Description | FL 100x/1.25/160/0.17 fluorescence free, oil, spring loaded |
| For Use With (Equipment) | Microscope models: FB69075 and FB69088 |
| Magnification Power | 100X |