Learn More
Leica Microsystems APO 0.63X Objective
Description
- Improved correction of chromatic aberration.
- Improved correction of spherical aberration.
- For S8 stereoscope microscopes.
Specifications
Specifications
| Type | Objective |
| Description | Working Distance: 100mm |
| For Use With (Equipment) | S8 Stereomicroscopes |