Learn More
Fisherbrand™ Achromatic Microscope Objective
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Description
- Achromatic microscope objective
- Accessory for microscope models: FB69019 and FB69020
Specifications
Specifications
| Type | Achromat Objective |
| Description | 40x/0.65/160/0.17 spring loaded |
| For Use With (Equipment) | Microscope models: FB69019 and FB69020 |
| Magnification Power | 40X |