Learn More
DuraLab Microscope Dust Cover for DR & DX Series
Product Code. 30280306
Change view
Click to view available options
Type:
Dust Cover
Unit Size:
Each
Product Code. Type unitSize
30280306 Dust Cover Each
1 options
Product Code. 30280306 Supplier DuraLab Supplier No. DRL0067
Only null left
Add to basket
Add to basket
Protect your microscope with this DuraLab dust cover designed for DR and DX series.
  • Maintains a clean, dust-free environment to extend the microscope's lifespan and ensure optimal performance
  • Dimensions: H 500 mm x W 400 mm
  • Compatible with most microscope brands-verify the size before purchasing
Specifications
Type Dust Cover
Compatible with DRL0040 - DuraLab DR-410 Advanced Achromatic Monocular Microscope, DRL0041 - DuraLab DR-420 Advanced Achromatic Binocular Microscope, DRL0042 - DuraLab DR-510 Advanced Semi-Plan Monocular Microscope, DRL0043 - DuraLab DR-520 Advanced Semi-Plan Binocular Microscope, DRL0044 - DuraLab DR-530 Advanced Semi-Plan Trinocular Microscope, DRL0045 - DuraLab DX-820 Pro Infinity Binocular Microscope, DRL0046 - DuraLab DX-830 Pro Infinity Trinocular Microscope