missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Microsart™ Media
Shop All Sartorius Products
Click to view available options
Without Additives:
No Cholesterol
No Sucrose
Unit Size:
Pack of 100
Description
- Ready-to-use agar media dish (prefilled with different agar media types) with active lid
- Sterile packaged and ready to use together with Microsart filter units
- Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
- Liftable interior lid provides easy access to select colonies after incubation for further analysis
Specifications
Specifications
| Product Type | Growth Medium |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Sterilization Method | Gamma-irradiated |
| With Additives | Essential and nonessential amino acids |
| Without Additives | No Cholesterol |
| For Use With (Application) | Target microorganisms: yeasts and molds |
| Formulation | Solid Agar Medium |
| Culture Type | Static Culture |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction