Learn More
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser
Product Code. 15953040 Shop All Sartorius Products
Change view
Click to view available options
Description:
2 VDC, 40VA
Lithium Battery (included)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15953040 Lithium Battery (included) Each
15963040 2 VDC, 40VA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15953040 Supplier Sartorius Stedim Biotech™ Supplier No. 16713BO
Only null left
Add to basket
Add to basket
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser facilitates procedures by automatically dispensing membrane filters.
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser facilitates procedures by automatically dispensing membrane filters.
  • Facilitates microbiological testing procedures.
  • Small footprint saves precious laminar flow hood space.
  • Dispenses membrane filters from their sterile packaging fully automatically at the touch of a button, hands-free, or by foot switch.
TRUSTED_SUSTAINABILITY
Specifications
Type Dispenser
For Use With (Application) Environmental/water testing, food and beverage testing, pharma/biotech
For Use With (Equipment) Membrane filters
Description Lithium Battery (included)
Weight (Metric) 2.4 kg
Dimensions (L x W x H) 233 mm × 172 mm × 230 mm
Dispensing Speed 0.5 sec.