missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Microsart™ AMP Extraction Kit

Product Code. 15494084 Shop All Sartorius Products
Click to view available options
Quantity:
50 Columns/Kit
Unit Size:
25 reactions
This item is not returnable. View return policy

Product Code. 15494084

Brand: Sartorius SMB952003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Microsart™ Mycoplasma AMP, Research quantitative detection kits.

  • Used for isolation of genomic DNA from different types of sample material
  • Compatible with Microsart™ Mycoplasma AMP, Research qPCR kits
  • Sensitive and robust detection of Mycoplasma within test samples

Compliance

EP 2.6.7

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Sample Type Cell Culture Supernatants
For Use With (Application) Mycoplasma Detection
Includes Buffers, collection tubes, sample storage tubes
Product Type Extraction Kit
Quantity 50 Columns/Kit
Sample Volume 200μL
Species Mycoplasma
Target DNA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.