missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Microplates for Fluorescence-based Assays

Product Code. 30274967
Change view
Click to view available options
No. of Wells:
96
384
Quantity:
10 Plates
30 Plates
Unit Size:
Pack of 10
Pack of 30
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Quantity Unit Size
30274967 96 30 Plates Pack of 30
15329413 96 10 Plates Pack of 10
18832653 384 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30274967 Supplier Invitrogen™ Supplier No. M33090
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
In Stock
Aggiungi al carrello
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.

Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).

Features of the microplates for fluorescence-based assays include:

  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
TRUSTED_SUSTAINABILITY

Specifica

Barcode No Barcode
Format Standard
Content And Storage Store at room temperature.
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 30-plates
Product Line Invitrogen
Color Clear Bottom, Black
Disposable Single-use
Product Type Microplates
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 200 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) Nucleic Acid Quantification
Quantity 30 Plates
Vedi altri risultati Mostra meno risultati

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.