Learn More
Ohaus™ Microplate Holder Kit
OHAUS™ Microplate Holder Kit for effortlessly mixing microtubes and microplates.
Brand: Ohaus™ 30400199
Additional Details : Weight : 1.10000kg
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Microplate Holder Kit | |
Plasitc | |
Micro Well Plate Kit with Retainer |