Learn More
Scientific Industries SI™ Microplate Foam Insert
Description
- Accommodates one standard footprint microplate.
- Fits into 152mm Platform attachment
- Included in the Multiple Sample Starter Set
- For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
Specifications
Specifications
| Material | High-density foam |
| For Use With (Equipment) | Vortex-Genie 2 family of mixers, Vortex-Genie Pulse |
| No. of Positions | 1 |
| Requires | Platform Attachment 15577315 |