Learn More
Restek Micropacked GC Column ShinCarbon ST 100/120 2m 1mmID 1/16"OD Silco General Config
Brand:  Restek 19808-800
Encompass_Preferred
Micropacked GC Column ShinCarbon ST 100/120 2m 1mmID 1/16"OD Silco General Config
Product Code. 16677236
  • £650.98 / Each