missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SST™ Striped Safety Tape

A striped, laminated vinyl safety tape that is designed for delineating and color coding specific equipment and areas within cleanrooms and controlled environments. Available in black/yellow, red/white, and black/white for differentiating marks.

Brand:  Micronova™ SST18-3BLYE

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13138525

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

For delineating and color coding specific equipment and areas

The Micronova SST™ Striped Safety Tape is a striped, pressure-sensitive tape with rubber adhesive that is ideal for marking fire equipment, exits, hazardous areas and first aid points within cleanrooms and controlled environments. The laminated vinyl is scuff resistant and holds up well to solvents, humidity and oil.

  • Laminated vinyl film with rubber adhesive
  • Cleanroom processed, pressure sensitive and scuff resistant
  • Designed for delineating and color coding in cleanrooms and controlled environments
  • Ideal for marking fire equipment, exits, hazardous areas and first aid points
  • Available in black/yellow, red/white, and black/white stripes
  • Holds up well to solvents, humidity and oil
  • Wound on a 3-inch plastic core

Order Info

Available in 1, 2 or 3 in. widths.

Specifications

Specifications

Not Autoclavable
Black, Yellow
Cleanroom Tape
Delineating and color-coding specific equipment and areas
16°C to 27°C
18 yd.
3 in.
Rubber
Vinyl
Non-sterile
60°F to 80°F
16.5 m x 76.2 mm (18 yd. x 3 in.)
16.5 m
76.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers