missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SST™ Striped Safety Tape

Product Code. 13108525 Shop All Micronova Products
Change view
Click to view available options
Color:
Black, White
Black, Yellow
Width (English):
2 in.
3 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Width (English) unitSize
13108525 Black, White 2 in. Each
13118525 Black, Yellow 2 in. Each
13138525 Black, Yellow 3 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13108525 Supplier Micronova™ Supplier No. SST182BLWH

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A striped, laminated vinyl safety tape that is designed for delineating and color coding specific equipment and areas within cleanrooms and controlled environments. Available in black/yellow, red/white, and black/white for differentiating marks.

For delineating and color coding specific equipment and areas

The Micronova SST™ Striped Safety Tape is a striped, pressure-sensitive tape with rubber adhesive that is ideal for marking fire equipment, exits, hazardous areas and first aid points within cleanrooms and controlled environments. The laminated vinyl is scuff resistant and holds up well to solvents, humidity and oil.

  • Laminated vinyl film with rubber adhesive
  • Cleanroom processed, pressure sensitive and scuff resistant
  • Designed for delineating and color coding in cleanrooms and controlled environments
  • Ideal for marking fire equipment, exits, hazardous areas and first aid points
  • Available in black/yellow, red/white, and black/white stripes
  • Holds up well to solvents, humidity and oil
  • Wound on a 3-inch plastic core

Order Info

Available in 1, 2 or 3 in. widths.

Specifications

Autoclavable Not Autoclavable
Adhesive Type Rubber
Coating Polylaminated Vinyl
Color Black, White
Finish Matte
Format Roll
Material Vinyl
Product Type Cleanroom Tape
Sterility Non-sterile
For Use With (Application) Delineating and color-coding specific equipment and areas
Temperature Range (English) 60°F to 80°F
Temperature Range (Metric) 16°C to 27°C
Dimensions (L x W) 16.5 m x 50.8 mm (18 yd. x 2 in.)
Length (English) 18 yd.
Length (Metric) 16.5 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.