missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Micromonospora viridifaciens NA Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Micromonospora viridifaciens Neuraminidase/Sialidase.
Specifications
Specifications
| Antigen | Micromonospora viridifaciens NA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Host Species | Rabbit |
| Immunogen | Recombinant Micromonospora viridifaciens Neuraminidase/Sialidase protein. |
| Purification Method | Protein A |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?