Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
Brand:  Epredia™ 524490
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12023059
  • £242.00 / Each
Estimated Shipment: 12-06-2024
to see stock.
Description
Description
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Specifications
Heat Extractor
HM 525