Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
GREENER_CHOICE
Product Code. 10144771
Change view
Click to view available options
Binding Property:
High Binding
Medium Binding
Surface Treatment:
Microlon 200
Microlon 600
Well Design:
Chimney Well
Standard Well
Unit Size:
Pack of 40
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Binding Property Surface Treatment Well Design unitSize
10144771 Medium Binding Microlon 200 Standard Well Pack of 40
10577091 High Binding Microlon 600 Standard Well Pack of 40
10043211 Medium Binding Microlon 600 Chimney Well Pack of 40
10680932 Medium Binding Microlon 200 Chimney Well Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10144771 Supplier Greiner Bio-One Supplier No. 655001
Only null left
Add to basket
Add to basket
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Tracebility from production through a defined LOT number system
  • Manufactured from crystal clear polystyrene
  • The footprint is compatible with automated systems
  • Well to well spacing : 9 mm
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Specifically treated to provide an incresed protein binding
  • Well profile: F-bottom / standard (flat bottom) 
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 200
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Standard Well