Learn More
Greiner Bio-One MICROLON™ LoBase μClear-Bottom 1536-well Microplate
Click to view available options
Color:
Black, Clear
Specifications
Specifications
| Format | Standard |
| Array | 32 x 48 |
| Color | Black, Clear |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Sterility | Sterile |
| Well Shape | Flat Bottom (F-well) |
| No. per Case | 60 |