missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates

Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Specifications
Specifications
| Breakapart | Single break |
| Binding Property | Medium Binding |
| Format | Strip Plate |
| Array | 8 x 12 |
| Packaging | Inner bags of 10 |
| Binding Type | Medium Binding |
| Color | Red, Clear Bottom |
| Product Type | 96-well Microplate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?