missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Service Plan
No
Warranty
No
Specifications
Specifications
| Breakapart | Single break |
| Binding Property | Medium Binding |
| Format | Strip Plate |
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Microlon |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?