missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MICROLON™ 200 12-well Elisa Strips
Description
- Polystyrene
- Medium binding
- 1 x 12 well
- Flat bottom strips for Plate Frame 701185
- Clear
Specifications
Specifications
| Color | Clear |
| Product Type | Elisa Strip |
| Material | Polystyrene |
| No. of Wells | 12 |
| Sterility | Non-sterile |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | ELISA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?