missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences MicroLiter 9 mm ScrewTin Autosampler Vial Caps with PTFE/Silicone Septa

Product Code. p-7237563 Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Pre-slit:
No
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 16498895

Brand: DWK Life Sciences 090030NT

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Available in multiple colors and package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material Polypropylene
Pre-slit No
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Type Screw Cap
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.