missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Microlink Cap

Product Code. 11718949 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
20-400
33-400
Unit Size:
Pack of 24
Pack of 48
This item is not returnable. View return policy

Product Code. 11718949

Brand: Wheaton™ 24025001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color White
For Use With (Application) Prolonged Period of Time Over Wide Temperature Range
Neck Finish 20-400
Product Type Microlink Cap
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.