Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System
Dilute/Dispense with better than 99% accuracy, independent of a liquid's viscosity, vapor pressure, and temperature. Microlab™ 600 Diluter/Dispenser System is a highly precise syringe pump with a graphical user interface designed to quickly and easily dilute and dispense volumes from 10μL to 50mL.
£251.00 - £10385.00
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
12341243
|
Hamilton™
ML625-DIL |
Dual Syringe Diluter | Advanced controller |
£10385.00
Each |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12365863
|
Hamilton™
61500-02 |
Basic to Advanced Controller Upgrade Kit | Advanced Manual, 2 GB SD Card, SD to USB Converter, Programmer Software CD |
£568.70
Each |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15329722
|
Hamilton™
61500-03 |
Custom Programmer Kit | Programmer Software CD with Programmer Manual, Application Programming Interface, and Example LabView, C#, and VB Programs |
£251.00
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12808495
|
Hamilton™
ML620-DIS |
Single Syringe Dispenser | Advanced controller |
£6645.00
Each |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12888485
|
Hamilton™
ML610-DIS |
Single Syringe Dispenser | Basic controller |
£5465.00
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16447820
|
Hamilton™
ML615-DIS |
Dual Syringe Dispenser | Basic controller |
£7080.00
Each |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15210157
|
Hamilton™
ML625-DIS |
Dual Syringe Dispenser | Advanced controller |
£7425.00
Each |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15136852
|
Hamilton™
ML615-DIL |
Dual Syringe Diluter | Basic controller |
£7730.00
1 unit |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Available as a single or dual syringe system
- High torque, precision stepper motors deliver positional accuracy
- Supports syringes from 10μL to 50mL (If no syringe(s) are selected at the time of the order, part numbers –DIS will ship with 1mL syringe(s) and numbers –CNT will ship with 10mL syringes.)
- Tubing diameters: 12 or 18 gauge
- Ethernet port for communication with controller or corporate network
- Printer Kit available
Syringe Pump Features
- High torque valve motors
- Precision syringe drives with 48,000 step resolution over 60 minutes
- Lighted power and prime buttons
- Independent left and right trigger ports
- Fanless heat vent
- 24V power input
- CAN Daisy Chain input/out
- RS-232 console port
- Power over Ethernet (PoE)
- TTL Input/Output
Dual Syringe Diluter
- Creates up to a 1:50,000 dilution in single step
- Reduces preparation time and wasted buffer
- Washes tubing between each sample
- Prints on standard, non-thermal 8 1/2 × 11 and A4 paper
- Interfaces with the free compliance software package that is available for Advanced Microlab™ devices
- Satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller
Dispenser: Sample spiking, Titration, Animal dosing (oral gavage), Reagent addition to chemical reactors, Flow chemistry, Manufacturing e.g. Mercury dispensing for light bulbs, Ink cartridge filling, Glue and epoxy dispensing, Nasal therapeutics (viscous), Contact lenses.
Diluter: Forensics blood alcohol analysis and urinalysis, minig assay labs metals detection, environmental analysis water and soils testing, oil analysis preventative maintenance, alcohol testing, beer and wine manufacturing.
NOTE: If no syringe(s) are selected at the time of the order, part numbers –DIS will ship with 1mL syringe(s) and numbers –CNT will ship with 10mL syringes.
Order Info
Microlab 600 Printer Kit sold separately
Specifications
Microlab 600 |