missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Microgard™ 2500 Plus PAPR Encapsulated Coveralls

Product Code. 15566564 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15566564 2X-Large Pack of 5
15546564 Large Pack of 5
15556564 X-Large Pack of 5
15536564 Medium Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15566564 Supplier Ansell™ Supplier No. WH25T0070106

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MICROGARD™ 2500 Plus PAPR Encapsulated Coveralls are designed for use with Powered Air Purifying Respirators (PAPRs) and certified to provide both respiratory and body protection from hazardous substances.

  • Fully encapsulated coverall, for use in combination with Scott Proflow PAPR fan unit, breathing hose and filters
  • Double elasticated cuffs, attached socks with boot over-flap.
  • Emergency rip cord attached to left chest shoulder and left back shoulder as worn
  • Achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
  • Anti-static according to EN 1149-5
  • Low linting for reduced risk of contamination in critical areas
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, Type 6 EN13034, EN14126, EN1073-2, EN1149-1
Color White
Hood Type Fully Encapsulated
Material Microporous Polypropylene Laminate
Size 2X-Large
Ankle Style Attached socks, Boot overflaps
Back Style Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
Cuff Style Double, Elastic
Description Emergency rip cord, attached socks with boot overflap
For Use With (Equipment) Scott Proflow 2SC/EX PAPR fan unit, breathing hoses and filters
No. of Pockets None
Product Type Protective coverall
Seam Type Stitched, Taped
Style Includes Emergency rip cord, attached socks with boot overflap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.