missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.
Brand: Agilent BioTek 7170011
Code : 04
Additional Details : Weight : 0.45360kg
Description
- Volume range 1 μL to 10 mL
- Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
- Dispense 1536 wells in 18 s.
- Cassettes can be autoclaved without recalibration
- Dispense up to eight reagents at one time
- Customizable dispense pattern allows skipped columns or rows
- Built-in priming trough
- Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
- Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput
Specifications
0.014 in. | |
For liquid handling | |
0.035 cm | |
Molded Polypropylene |
5 to 2500 μL | |
Medium Volume Cassette | |
5 μL | |
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |